  • Search Results
  • Multivalent Flexible Nanogels Exhibit Broad-Spectrum Antiviral Activity by Blocking Virus Entry.
Multivalent Flexible Nanogels Exhibit Broad-Spectrum Antiviral Activity by Blocking Virus Entry.
ACS nano (2018-06-13)
Pradip Dey, Tobias Bergmann, Jose Luis Cuellar-Camacho, Svenja Ehrmann, Mohammad Suman Chowdhury, Minze Zhang, Ismail Dahmani, Rainer Haag, Walid Azab
ABSTRACT
The entry process of viruses into host cells is complex and involves stable but transient multivalent interactions with different cell surface receptors. The initial contact of several viruses begins with attachment to heparan sulfate (HS) proteoglycans on the cell surface, which results in a cascade of events that end up with virus entry. The development of antiviral agents based on multivalent interactions to shield virus particles and block initial interactions with cellular receptors has attracted attention in antiviral research. Here, we designed nanogels with different degrees of flexibility based on dendritic polyglycerol sulfate to mimic cellular HS. The designed nanogels are nontoxic and broad-spectrum, can multivalently interact with viral glycoproteins, shield virus surfaces, and efficiently block infection. We also visualized virus-nanogel interactions as well as the uptake of nanogels by the cells through clathrin-mediated endocytosis using confocal microscopy. As many human viruses attach to the cells through HS moieties, we introduce our flexible nanogels as robust inhibitors for these viruses.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Methyl cellulose, viscosity: 400 cP
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 70,000, (FITC:Glucose = 1:250)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)
Sigma-Aldrich
Methyl-β-cyclodextrin, average Mn 1310
Sigma-Aldrich
Dynasore hydrate
Sigma-Aldrich
5-(N-Ethyl-N-isopropyl)amiloride