Merck
  • TDP-43 enhances translation of specific mRNAs linked to neurodegenerative disease.
TDP-43 enhances translation of specific mRNAs linked to neurodegenerative disease.
Nucleic acids research (2018-10-26)
Nagammal Neelagandan, Giorgio Gonnella, Stefan Dang, Philipp C Janiesch, Katharine K Miller, Katrin Küchler, Rita F Marques, Daniela Indenbirken, Malik Alawi, Adam Grundhoff, Stefan Kurtz, Kent E Duncan
ABSTRACT
The RNA-binding protein TDP-43 is heavily implicated in neurodegenerative disease. Numerous patient mutations in TARDBP, the gene encoding TDP-43, combined with data from animal and cell-based models, imply that altered RNA regulation by TDP-43 causes Amyotrophic Lateral Sclerosis and Frontotemporal Dementia. However, underlying mechanisms remain unresolved. Increased cytoplasmic TDP-43 levels in diseased neurons suggest a possible role in this cellular compartment. Here, we examined the impact on translation of overexpressing human TDP-43 and the TDP-43A315T patient mutant protein in motor neuron-like cells and primary cultures of cortical neurons. In motor-neuron like cells, TDP-43 associates with ribosomes without significantly affecting global translation. However, ribosome profiling and additional assays revealed enhanced translation and direct binding of Camta1, Mig12, and Dennd4a mRNAs. Overexpressing either wild-type TDP-43 or TDP-43A315T stimulated translation of Camta1 and Mig12 mRNAs via their 5'UTRs and increased CAMTA1 and MIG12 protein levels. In contrast, translational enhancement of Dennd4a mRNA required a specific 3'UTR region and was specifically observed with the TDP-43A315T patient mutant allele. Our data reveal that TDP-43 can function as an mRNA-specific translational enhancer. Moreover, since CAMTA1 and DENND4A are linked to neurodegeneration, they suggest that this function could contribute to disease.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-MID1IP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)