CH
EN
Applications
Products
Services
Support
Home
Search Results
Production of a gonadotropin-releasing hormone 2 receptor knockdown (GNRHR2 KD) swine line
Production of a gonadotropin-releasing hormone 2 receptor knockdown (GNRHR2 KD) swine line
Transgenic research (2017)
Desaulniers AT, et al.