Merck
Use of inhibitors in the study of MAP kinases.
Methods in molecular biology (Clifton, N.J.) (2010-09-03)
Kimberly Burkhard, Paul Shapiro
ABSTRACT
The mitogen-activated protein (MAP) kinases are ubiquitous intracellular signaling proteins that respond to a variety of extracellular signals and regulate most cellular functions including proliferation, apoptosis, migration, differentiation, and secretion. The four major MAP kinase family members, which include the ERK1/2, JNK, p38, and ERK5 proteins, coordinate cellular responses by phosphorylating and regulating the activity of dozens of substrate proteins involved in transcription, translation, and changes in cellular architecture. Uncontrolled activation of the MAP kinases has been implicated in the initiation and progression of a variety of cancers and inflammatory disorders. As such, the ability to manipulate the activity of MAP kinase proteins with specific pharmacological inhibitors has received much attention as research tools for understanding basic mechanisms of cellular functions and for clinical tools to treat diseases. A variety of pharmacological inhibitors have been developed to selectively block MAP kinases directly or indirectly through targeting upstream regulators. This chapter will provide an overview of some of the current inhibitors that target MAP kinase signaling pathways and provide methodology on how to use selective MAP kinase inhibitors and immunoblotting techniques to monitor and quantify phosphorylation of MAP kinase substrates.
Product Description
Supelco
N-Nitrosodi-n-butylamine, analytical standard
Sigma-Aldrich
U0126, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC50 = 72 nM) and MEK2 (IC50 = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.
Sigma-Aldrich
Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody, Mouse monoclonal, clone MAPK-YT, purified from hybridoma cell culture
Sigma-Aldrich
Anti-p38 MAP Kinase (341-360) Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
SB 203580, SB 203580, CAS 152121-47-6, is a highly specific, potent, cell-permeable, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC50 = 34 nM in vitro, 600 nM in cells).
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, phosphoSer31, Chemicon®, from rabbit