Merck
  • DEET and other repellents are inhibitors of mosquito odorant receptors for oviposition attractants.
DEET and other repellents are inhibitors of mosquito odorant receptors for oviposition attractants.
Insect biochemistry and molecular biology (2019-08-26)
Pingxi Xu, Fangfang Zeng, Robert H Bedoukian, Walter S Leal
ABSTRACT
In addition to its primary function as an insect repellent, DEET has many "off-label" properties, including a deterrent effect on the attraction of gravid female mosquitoes. DEET negatively affects oviposition sites. While deorphanizing odorant receptors (ORs) using the Xenopus oocyte recording system, we have previously observed that DEET generated outward (inhibitory) currents on ORs sensitive to oviposition attractants. Here, we systematically investigated these inhibitory currents. We recorded dose-dependent outward currents elicited by DEET and other repellents on ORs from Culex quinquefasciatus, Aedes aegypti, and Anopheles gambiae. Similar responses were observed with other plant-derived and plant-inspired compounds, including methyl jasmonate and methyl dihydrojasmolate. Inward (regular) currents elicited by skatole upon activation of CquiOR21 were modulated when this oviposition attractant was coapplied with a repellent. Compounds that generate outward currents in ORs sensitive to oviposition attractants elicited inward currents in a DEET-sensitive receptor, CquiOR136. The best ligand for this receptor, methyl dihydrojasmolate, showed repellency activity but was not as strong as DEET in our test protocol.
Product Description
Stainless steel 304 syringe needle, gauge 16, L 6 in.
Pricing and availability is not currently available.