Merck
  • Clarin-2 is essential for hearing by maintaining stereocilia integrity and function.
Clarin-2 is essential for hearing by maintaining stereocilia integrity and function.
EMBO molecular medicine (2019-08-27)
Lucy A Dunbar, Pranav Patni, Carlos Aguilar, Philomena Mburu, Laura Corns, Helena Rr Wells, Sedigheh Delmaghani, Andrew Parker, Stuart Johnson, Debbie Williams, Christopher T Esapa, Michelle M Simon, Lauren Chessum, Sherylanne Newton, Joanne Dorning, Prashanthini Jeyarajan, Susan Morse, Andrea Lelli, Gemma F Codner, Thibault Peineau, Suhasini R Gopal, Kumar N Alagramam, Ronna Hertzano, Didier Dulon, Sara Wells, Frances M Williams, Christine Petit, Sally J Dawson, Steve Dm Brown, Walter Marcotti, Aziz El-Amraoui, Michael R Bowl
ABSTRACT
Hearing relies on mechanically gated ion channels present in the actin-rich stereocilia bundles at the apical surface of cochlear hair cells. Our knowledge of the mechanisms underlying the formation and maintenance of the sound-receptive structure is limited. Utilizing a large-scale forward genetic screen in mice, genome mapping and gene complementation tests, we identified Clrn2 as a new deafness gene. The Clrn2clarinet/clarinet mice (p.Trp4* mutation) exhibit a progressive, early-onset hearing loss, with no overt retinal deficits. Utilizing data from the UK Biobank study, we could show that CLRN2 is involved in human non-syndromic progressive hearing loss. Our in-depth morphological, molecular and functional investigations establish that while it is not required for initial formation of cochlear sensory hair cell stereocilia bundles, clarin-2 is critical for maintaining normal bundle integrity and functioning. In the differentiating hair bundles, lack of clarin-2 leads to loss of mechano-electrical transduction, followed by selective progressive loss of the transducing stereocilia. Together, our findings demonstrate a key role for clarin-2 in mammalian hearing, providing insights into the interplay between mechano-electrical transduction and stereocilia maintenance.
Sigma-Aldrich
Anti-Opsin Antibody, blue, Chemicon®, from rabbit
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Iba1/AIF1 Antibody, clone 20A12.1, from mouse