Merck
  • DIA-NN: neural networks and interference correction enable deep proteome coverage in high throughput.
DIA-NN: neural networks and interference correction enable deep proteome coverage in high throughput.
Nature methods (2019-11-27)
Vadim Demichev, Christoph B Messner, Spyros I Vernardis, Kathryn S Lilley, Markus Ralser
ABSTRACT
We present an easy-to-use integrated software suite, DIA-NN, that exploits deep neural networks and new quantification and signal correction strategies for the processing of data-independent acquisition (DIA) proteomics experiments. DIA-NN improves the identification and quantification performance in conventional DIA proteomic applications, and is particularly beneficial for high-throughput applications, as it is fast and enables deep and confident proteome coverage when used in combination with fast chromatographic methods.
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, Molecular Biology, ≥99.5% (RT)
Sigma-Aldrich
Yeast Nitrogen Base Without Amino Acids, Yeast classification medium used for selecting yeasts based on amino acid and carbohydrate requirements
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)