Merck
  • Renal Purge of Hemolymphatic Lipids Prevents the Accumulation of ROS-Induced Inflammatory Oxidized Lipids and Protects Drosophila from Tissue Damage.
Renal Purge of Hemolymphatic Lipids Prevents the Accumulation of ROS-Induced Inflammatory Oxidized Lipids and Protects Drosophila from Tissue Damage.
Immunity (2020-02-23)
Xiaoxue Li, Samuel Rommelaere, Shu Kondo, Bruno Lemaitre
ABSTRACT
Animals require complex metabolic and physiological adaptations to maintain the function of vital organs in response to environmental stresses and infection. Here, we found that infection or injury in Drosophila induced the excretion of hemolymphatic lipids by Malpighian tubules, the insect kidney. This lipid purge was mediated by a stress-induced lipid-binding protein, Materazzi, which was enriched in Malpighian tubules. Flies lacking materazzi had higher hemolymph concentrations of reactive oxygen species (ROS) and increased lipid peroxidation. These flies also displayed Malpighian tubule dysfunction and were susceptible to infections and environmental stress. Feeding flies with antioxidants rescued the materazzi phenotype, indicating that the main role of Materazzi is to protect the organism from damage caused by stress-induced ROS. Our findings suggest that purging hemolymphatic lipids presents a physiological adaptation to protect host tissues from excessive ROS during immune and stress responses, a process that is likely to apply to other organisms.
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol
Sigma-Aldrich
Glycerol Standard Solution, 2.5 mg/ml equivalent triolein concentration
Supelco
Dimethyl sulfone, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
1,3-Diolein, ≥99% (GC)
Sigma-Aldrich
Phospholipid Assay Kit, sufficient for 100 colorimetric or fluorometric tests
Sigma-Aldrich
Fluorimetric Hydrogen Peroxide Assay Kit, sufficient for 500 fluorometric tests (red fluorescence)
Sigma-Aldrich
DAPI, dilactate, ≥98% (HPLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Supelco
Glucose (HK) Assay Kit, sufficient for 20 assays
Sigma-Aldrich
Triglyceride Reagent, used for quantitative enzymatic determination of triglyerides