  • Search Results
  • Targeting downstream subcellular YAP activity as a function of matrix stiffness with Verteporfin-encapsulated chitosan microsphere attenuates osteoarthritis.
Targeting downstream subcellular YAP activity as a function of matrix stiffness with Verteporfin-encapsulated chitosan microsphere attenuates osteoarthritis.
Biomaterials (2020-01-10)
Xianzhu Zhang, Dandan Cai, Feifei Zhou, Jie Yu, Xinyu Wu, Dongsheng Yu, Yiwei Zou, Yi Hong, Chunhui Yuan, Yishan Chen, Zongyou Pan, Varitsara Bunpetch, Heng Sun, Chengrui An, Toh Yi-Chin, Hongwei Ouyang, Shufang Zhang
ABSTRACT
Changes in the stiffness of chondrocyte extracellular matrix (ECM) are involved in the pathological progression of osteoarthritis (OA). However, the downstream responses of cartilage ECM stiffness are still unclear. YAP (Yes-associated protein) has been extensively studied as a mechanotransducer, we thus hypothesized that by targeting the downstream molecule activity of ECM stiffness could maintain chondrocyte phenotype and prevent cartilage degeneration in OA. Here, we showed that human cartilage matrix stiffened during pathological progression of OA, and the chondrocyte YAP activity was associated with ECM stiffness. We then mimicked the physiological and pathological stiffness of human cartilage by using PDMS-based substrates, and found that YAP was activated in chondrocytes seeded on stiff substrate, gradually losing their phenotype. In addition, it was observed that YAP was also significantly activated in mice OA development, and conditional knockout (cKO) of YAP in mice preserved collagen II expression and protected cartilage from degeneration in the OA model. Furthermore, intra-articular injection of YAP-selective inhibitor, Verteporfin, significantly maintained cartilage homeostasis in mice OA model. This study indicates that the application of mechanotransducer-targeted drugs could be a potential therapeutic approach for cartilage repair in OA.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Leptomycin B solution from Streptomyces sp., ≥95% (HPLC), Supplied in methanol: water (7:3)
Sigma-Aldrich
Verteporfin, ≥94% (HPLC)