Merck
  • Phospholipase D-dependent mTOR complex 1 (mTORC1) activation by glutamine.
Phospholipase D-dependent mTOR complex 1 (mTORC1) activation by glutamine.
The Journal of biological chemistry (2018-09-09)
Elyssa Bernfeld, Deepak Menon, Vishaldeep Vaghela, Ismat Zerin, Promie Faruque, Maria A Frias, David A Foster
ABSTRACT
Glutamine is a key nutrient required for sustaining cell proliferation, contributing to nucleotide, protein, and lipid synthesis. The mTOR complex 1 (mTORC1) is a highly conserved protein complex that acts as a sensor of nutrients, relaying signals for the shift from catabolic to anabolic metabolism. Although glutamine plays an important role in mTORC1 activation, the mechanism is not clear. Here we describe a leucine- and Rag-independent mechanism of mTORC1 activation by glutamine that depends on phospholipase D and the production of phosphatidic acid, which is required for the stability and activity of mTORC1. The phospholipase D-dependent activation of mTORC1 by glutamine depended on the GTPases ADP ribosylation factor 1 (Arf1), RalA, and Rheb. Glutamine deprivation could be rescued by α-ketoglutarate, a downstream metabolite of glutamine. This mechanism represents a distinct nutrient input to mTORC1 that is independent of Rag GTPases and leucine.
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture