Merck
  • L ARP7 Is a BRCA1 Ubiquitinase Substrate and Regulates Genome Stability and Tumorigenesis.
L ARP7 Is a BRCA1 Ubiquitinase Substrate and Regulates Genome Stability and Tumorigenesis.
Cell reports (2020-07-30)
Fang Zhang, Pengyi Yan, Huijing Yu, Huangying Le, Zixuan Li, Jiahuan Chen, Xiaodong Liang, Shiyan Wang, Weiting Wei, Li Liu, Yan Zhang, Xing Ji, Anyong Xie, Wantao Chen, Zeguang Han, William T Pu, Sun Chen, Yingwei Chen, Kun Sun, Baoxue Ge, Bing Zhang
ABSTRACT
Attenuated DNA repair leads to genomic instability and tumorigenesis. BRCA1/BARD1 are the best-known tumor suppressors that promote homology recombination (HR) and arrest cell cycle. However, it remains ambiguous whether and how their E3 ligase activity regulates HR. Here, we demonstrate that upon genotoxic stress, BRCA1 together with BARD1 catalyzes the K48 polyubiquitination on LARP7, a 7SK RNA binding protein known to control RNAPII pausing, and thereby degrades it through the 26S ubiquitin-proteasome pathway. Depleting LARP7 suppresses the expression of CDK1 complex, arrests the cell at the G2/M DNA damage checkpoint, and reduces BRCA2 phosphorylation, which thereby facilitates RAD51 recruitment to damaged DNA to enhance HR. Importantly, LARP7 depletion observed in breast cancer patients leads to chemoradiotherapy resistance both in vitro and in vivo. Altogether, this study unveils a mechanism by which BRCA1/BARD1 control HR and cell cycle, and highlights LARP7 as a potential target for cancer prevention and therapy.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-Cyclin B1 Antibody, clone GNS3 (8A5D12), clone GNS3 (8A5D12), Upstate®, from mouse
Sigma-Aldrich
Anti-BRCA2 (Ab-1) Mouse mAb (2B), liquid, clone 2B, Calbiochem®
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)
Sigma-Aldrich
Anti-BRCA1 (Ab-1) Mouse mAb (MS110), liquid, clone MS110, Calbiochem®
Sigma-Aldrich
Anti-ATM phosphoSer1981 Antibody, clone 10H11.E12, clone 10H11.E12, Chemicon®, from mouse
Sigma-Aldrich
Anti-phospho-BRCA2 (Ser3291) Antibody, from rabbit, purified by affinity chromatography