Merck
  • Gnathodiaphyseal dysplasia is not recapitulated in a respective mouse model carrying a mutation of the Ano5 gene.
Gnathodiaphyseal dysplasia is not recapitulated in a respective mouse model carrying a mutation of the Ano5 gene.
Bone reports (2020-05-27)
Tim Rolvien, Osman Avci, Simon von Kroge, Till Koehne, Stefan Selbert, Stephan Sonntag, Doron Shmerling, Uwe Kornak, Ralf Oheim, Michael Amling, Thorsten Schinke, Timur Alexander Yorgan
ABSTRACT
Mutations in the gene ANO5, encoding for the transmembrane protein Anoctamin 5 (Ano5), have been identified to cause gnathodiaphyseal dysplasia (GDD) in humans, a skeletal disorder characterized by sclerosis of tubular bones, increased fracture risk and fibro-osseous lesions of the jawbones. To better understand the pathomechanism of GDD we have generated via Crispr/CAS9 gene editing a mouse model harboring the murine equivalent (Ano5 p.T491F) of a GDD-causing ANO5 mutation identified in a previously reported patient. Skeletal phenotyping by contact radiography, μCT and undecalcified histomorphometry was performed in male mice, heterozygous and homozygous for the mutation, at the ages of 12 and 24 weeks. These mice did not display alterations of skeletal microarchitecture or mandible morphology. The results were confirmed in female mice and animals derived from a second, independent clone. Finally, no skeletal phenotype was observed in mice lacking ~40% of their Ano5 gene due to a frameshift mutation. Therefore, our results indicate that Ano5 is dispensable for bone homeostasis in mice, at least under unchallenged conditions, and that these animals may not present the most adequate model to study the physiological role of Anoctamin 5.
Sigma-Aldrich
AST Activity Assay Kit, sufficient for 100 colorimetric tests
Sigma-Aldrich
Brain Natriuretic Peptide EIA Kit, for serum, plasma, culture supernatant and cell lysates
Sigma-Aldrich
ALT Activity Assay, sufficient for 100 colorimetric or fluorometric tests