  • Search Results
  • Identification, Characterization, and Heritability of Murine Metastable Epialleles: Implications for Non-genetic Inheritance.
Identification, Characterization, and Heritability of Murine Metastable Epialleles: Implications for Non-genetic Inheritance.
Cell (2018-11-21)
Anastasiya Kazachenka, Tessa M Bertozzi, Marcela K Sjoberg-Herrera, Nic Walker, Joseph Gardner, Richard Gunning, Elena Pahita, Sarah Adams, David Adams, Anne C Ferguson-Smith
ABSTRACT
Generally repressed by epigenetic mechanisms, retrotransposons represent around 40% of the murine genome. At the Agouti viable yellow (Avy) locus, an endogenous retrovirus (ERV) of the intracisternal A particle (IAP) class retrotransposed upstream of the agouti coat-color locus, providing an alternative promoter that is variably DNA methylated in genetically identical individuals. This results in variable expressivity of coat color that is inherited transgenerationally. Here, a systematic genome-wide screen identifies multiple C57BL/6J murine IAPs with Avy epigenetic properties. Each exhibits a stable methylation state within an individual but varies between individuals. Only in rare instances do they act as promoters controlling adjacent gene expression. Their methylation state is locus-specific within an individual, and their flanking regions are enriched for CTCF. Variably methylated IAPs are reprogrammed after fertilization and re-established as variable loci in the next generation, indicating reconstruction of metastable epigenetic states and challenging the generalizability of non-genetic inheritance at these regions.
Sigma-Aldrich
Imprint® DNA Modification Kit, For bisulfite DNA conversion & purification