Merck
  • Spatiotemporal expression of aquaporin 9 is critical for the antral growth of mouse ovarian follicles†.
Spatiotemporal expression of aquaporin 9 is critical for the antral growth of mouse ovarian follicles†.
Biology of reproduction (2020-06-25)
Sungeun Lee, Hee-Gyoo Kang, Chongsuk Ryou, Yong-Pil Cheon
ABSTRACT
Although a few aquaporins (AQPs) expressed in granulosa cells have been postulated to mediate fluid passage into the antrum, the specific expression of AQPs in different follicle cell types and stages and their roles have not been evaluated extensively. The spatiotemporal expression of aquaporin (Aqp) 7, 8, and 9 and the functional roles of Aqp9 in antral growth and ovulation were examined using a superovulation model and 3-dimensional follicle culture. Aqp9 was expressed at a high level in the rapid growth phase (24-48 h post equine chorionic gonadotropin (eCG) for superovulation induction) compared to Aqp7 (after human chorionic gonadotropin (hCG)) and Aqp8 (8-24 h post eCG and 24 h post hCG). A dramatic increase in the expression and localization of Aqp9 mRNA in theca cells was observed, as evaluated using quantitative reverse transcription-polymerase (RT-PCR) coupled with laser capture microdissection and immunohistochemistry. AQP9 was located primarily on the theca cells of the tertiary and preovulatory follicles but not on the ovulated follicles. In phloretin-treated mice, the diameter of the preovulatory follicles and the number of ovulated oocytes decreased. Consistent with these findings, knocking down Aqp9 expression with an Aqp9 siRNA inhibited follicle growth (0.28:1 = siRNA:control) and decreased the number of ovulated follicles (0.36:1 = siRNA:control) during in vitro growth and ovulation induction. Based on these results, the expression of AQPs is under the control of the physiological status, and AQP9 expression in theca during folliculogenesis is required for antral growth and ovulation in a tissue-specific and stage-dependent manner.
Sigma-Aldrich
Follicle Stimulating Hormone from human pituitary, ~7,000 IU/mg (powder)
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Transferrin human, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phloretin, ≥99%
Sigma-Aldrich
Chorionic gonadotropin human, lyophilized powder, vial of ~2,500 IU