Merck
Deletions in CWH43 cause idiopathic normal pressure hydrocephalus.
EMBO molecular medicine (2021-01-19)
Hong Wei Yang, Semin Lee, Dejun Yang, Huijun Dai, Yan Zhang, Lei Han, Sijun Zhao, Shuo Zhang, Yan Ma, Marciana F Johnson, Anna K Rattray, Tatyana A Johnson, George Wang, Shaokuan Zheng, Rona S Carroll, Peter J Park, Mark D Johnson
ABSTRACT
Idiopathic normal pressure hydrocephalus (iNPH) is a neurological disorder that occurs in about 1% of individuals over age 60 and is characterized by enlarged cerebral ventricles, gait difficulty, incontinence, and cognitive decline. The cause and pathophysiology of iNPH are largely unknown. We performed whole exome sequencing of DNA obtained from 53 unrelated iNPH patients. Two recurrent heterozygous loss of function deletions in CWH43 were observed in 15% of iNPH patients and were significantly enriched 6.6-fold and 2.7-fold, respectively, when compared to the general population. Cwh43 modifies the lipid anchor of glycosylphosphatidylinositol-anchored proteins. Mice heterozygous for CWH43 deletion appeared grossly normal but displayed hydrocephalus, gait and balance abnormalities, decreased numbers of ependymal cilia, and decreased localization of glycosylphosphatidylinositol-anchored proteins to the apical surfaces of choroid plexus and ependymal cells. Our findings provide novel mechanistic insights into the origins of iNPH and demonstrate that it represents a distinct disease entity.
Sigma-Aldrich
Anti-CWH43 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.