  • Search Results
  • Phytochemical Profile, Antioxidant, Cardioprotective and Nephroprotective Activity of Romanian Chicory Extract.
Phytochemical Profile, Antioxidant, Cardioprotective and Nephroprotective Activity of Romanian Chicory Extract.
Plants (Basel, Switzerland) (2021-01-06)
Alexandra Epure, Alina E E Pârvu, Laurian Vlase, Daniela Benedec, Daniela Hanganu, Ana-Maria Gheldiu, Vlad Al Toma, Ilioara Oniga
ABSTRACT
The present study analyzed the methanol extract and tincture obtained from the spontaneous Romanian Cichorium intybus species, in order to evaluate polyphenols content and some biological properties. Chromatographic and spectrophotometric methods were used for the analysis of polyphenols and the antioxidant capacity was assessed in vitro with DPPH● (2,2-diphenyl-picrylhydrazil) and FRAP (ferric-reducing antioxidant power) tests. The cardio-protective effects of Cichorii herba tincture on myocardial ischemia induced by isoprenaline and nephroprotection on renal failure induced by gentamicin were evaluated on rats. Also, aspartate aminotrasferase (AST), alanine aminotransferase (ALT), creatine kinase-MB (CK-MB) and creatinine clearance (CrCl) were measured. The antioxidant effect was evaluated by determining total oxidative stress (TOS), oxidative stress index (OSI, total antioxidant capacity (TAC), malondyaldehide (MDA), total thiols (SH) and total nitrites and nitrates (NOx). Cichoric acid was the main polyphenolic compound. The extracts had moderate in vitro antioxidant activity but the in vivo antioxidant and anti-inflammatory effects were significant and associated with myocardial and renal dysfunction improvement. The results were attributed to the content of polyphenols in the extracts, for which reason C. intybus may be considered an important raw material for pharmaceuticals formulations recommended in the prevention or treatment of heart or kidney diseases.
Sigma-Aldrich
Caftaric acid, ≥97.0%
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Rutin, phyproof® Reference Substance