  • Search Results
  • Genes responsive to rapamycin and serum deprivation are clustered on chromosomes and undergo reorganization within local chromatin environments.
Genes responsive to rapamycin and serum deprivation are clustered on chromosomes and undergo reorganization within local chromatin environments.
Biochemistry and cell biology = Biochimie et biologie cellulaire (2019-09-04)
Zachery R Belak, Joshua A Pickering, Zoe E Gillespie, Gerald Audette, Mark Eramian, Jennifer A Mitchell, Joanna M Bridger, Anthony Kusalik, Christopher H Eskiw
ABSTRACT
We previously demonstrated that genome reorganization, through chromosome territory repositioning, occurs concurrently with significant changes in gene expression in normal primary human fibroblasts treated with the drug rapamycin, or stimulated into quiescence. Although these events occurred concomitantly, it is unclear how specific changes in gene expression relate to reorganization of the genome at higher resolution. We used computational analyses, genome organization assays, and microscopy, to investigate the relationship between chromosome territory positioning and gene expression. We determined that despite relocation of chromosome territories, there was no substantial bias in the proportion of genes changing expression on any one chromosome, including chromosomes 10 and 18. Computational analyses identified that clusters of serum deprivation and rapamycin-responsive genes along the linear extent of chromosomes. Chromosome conformation capture (3C) analysis demonstrated the strengthening or loss of specific long-range chromatin interactions in response to rapamycin and quiescence induction, including a cluster of genes containing Interleukin-8 and several chemokine genes on chromosome 4. We further observed that the LIF gene, which is highly induced upon rapamycin treatment, strengthened interactions with up- and down-stream intergenic regions. Our findings indicate that the repositioning of chromosome territories in response to cell stimuli, this does not reflect gene expression changes occurring within physically clustered groups of genes.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution