Merck
  • Age-related increase in caveolin-1 expression facilitates cell-to-cell transmission of α-synuclein in neurons.
Age-related increase in caveolin-1 expression facilitates cell-to-cell transmission of α-synuclein in neurons.
Molecular brain (2021-07-30)
Tae-Young Ha, Yu Ree Choi, Hye Rin Noh, Seon-Heui Cha, Jae-Bong Kim, Sang Myun Park
ABSTRACT
Parkinson's disease (PD) is the second most prevalent neurodegenerative disease, with aging being considered the greatest risk factor for developing PD. Caveolin-1 (Cav-1) is known to participate in the aging process. Recent evidence indicates that prion-like propagation of misfolded α-synuclein (α-syn) released from neurons to neighboring neurons plays an important role in PD progression. In the present study, we demonstrated that cav-1 expression in the brain increased with age, and considerably increased in the brain of A53T α-syn transgenic mice. Cav-1 overexpression facilitated the uptake of α-syn into neurons and formation of additional Lewy body-like inclusion bodies, phosphorylation of cav-1 at tyrosine 14 was found to be crucial for this process. This study demonstrates the relationship between age and α-syn spread and will facilitate our understanding of the molecular mechanism of the cell-to-cell transmission of α-syn.
Sigma-Aldrich
Src Inhibitor-1, ≥98% (HPLC)
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)