Merck
Single-Cell Western Blotting.
Methods in molecular biology (Clifton, N.J.) (2015-11-07)
Elly Sinkala, Amy E Herr
ABSTRACT
Little headway has been made in single cell protein analysis, aside from tools that rely solely on antibody-probe based detection (i.e., flow cytometry, immunocytochemistry), which are limited by low specificity and multiplexing capabilities. To address these protein analysis gaps, we have introduced a single-cell western blot (scWestern). The protein assay is capable of highly specific analysis by coupling antibody-based detection with a polyacrylamide gel electrophoresis (PAGE) protein separation. Cells are settled via gravity into polyacrylamide (PA) microwells, chemically lysed in the wells, and then subjected to PAGE through the walls of the microwells and into the surrounding PA gel. Over a thousand single-cell separations are performed simultaneously, and multiple protein targets of interest are investigated. After PAGE separation, photo-immobilization of all proteins to the gel allows for antibody probing and lends to the archival quality of the scWestern assay where new proteins targets can be investigated months after the initial separations are performed.
Sigma-Aldrich
Saponin, Molecular Biology, used as non-ionic surfactant
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, suitable for electrophoresis, suitable for acrylamide photopolymerization, ≥70%, powder
Sigma-Aldrich
Anti-Nestin Antibody, clone rat-401, clone rat-401, Chemicon®, from mouse
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Ammonium persulfate, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Poly-L-ornithine hydrobromide, mol wt 30,000-70,000
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Anti-β-Tubulin III (neuronal) antibody, Mouse monoclonal, ~1.0 mg/mL, clone 2G10, purified from hybridoma cell culture