Merck
  • High-fat diet-activated fatty acid oxidation mediates intestinal stemness and tumorigenicity.
High-fat diet-activated fatty acid oxidation mediates intestinal stemness and tumorigenicity.
Cell reports (2021-06-10)
Miyeko D Mana, Amanda M Hussey, Constantine N Tzouanas, Shinya Imada, Yesenia Barrera Millan, Dorukhan Bahceci, Dominic R Saiz, Anna T Webb, Caroline A Lewis, Peter Carmeliet, Maria M Mihaylova, Alex K Shalek, Ömer H Yilmaz
ABSTRACT
Obesity is an established risk factor for cancer in many tissues. In the mammalian intestine, a pro-obesity high-fat diet (HFD) promotes regeneration and tumorigenesis by enhancing intestinal stem cell (ISC) numbers, proliferation, and function. Although PPAR (peroxisome proliferator-activated receptor) nuclear receptor activity has been proposed to facilitate these effects, their exact role is unclear. Here we find that, in loss-of-function in vivo models, PPARα and PPARδ contribute to the HFD response in ISCs. Mechanistically, both PPARs do so by robustly inducing a downstream fatty acid oxidation (FAO) metabolic program. Pharmacologic and genetic disruption of CPT1A (the rate-controlling enzyme of mitochondrial FAO) blunts the HFD phenotype in ISCs. Furthermore, inhibition of CPT1A dampens the pro-tumorigenic consequences of a HFD on early tumor incidence and progression. These findings demonstrate that inhibition of a HFD-activated FAO program creates a therapeutic opportunity to counter the effects of a HFD on ISCs and intestinal tumorigenesis.
Sigma-Aldrich
TRI Reagent®
Sigma-Aldrich
Sodium octanoate, ≥99% (capillary GC)
Sigma-Aldrich
N-Acetyl-L-cysteine, suitable for cell culture, BioReagent
Sigma-Aldrich
(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
BrdU (5-Bromo-2′-deoxyuridine), Used for the study of DNA synthesis where it is incorporated into DNA in place of thymidine, BrdU can be used in conjunction with anti-BrdU for immunocytochemical analysis of cell proliferation.
Sigma-Aldrich
GW501516, ≥98% (HPLC)
Sigma-Aldrich
Sodium L-lactate, ~98%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)