Merck
  • AXL Inhibition in Macrophages Stimulates Host-versus-Leukemia Immunity and Eradicates Naïve and Treatment-Resistant Leukemia.
AXL Inhibition in Macrophages Stimulates Host-versus-Leukemia Immunity and Eradicates Naïve and Treatment-Resistant Leukemia.
Cancer discovery (2021-06-10)
Irene Tirado-Gonzalez, Arnaud Descot, Devona Soetopo, Aleksandra Nevmerzhitskaya, Alexander Schäffer, Ivan-Maximilano Kur, Ewelina Czlonka, Carolin Wachtel, Ioanna Tsoukala, Luise Müller, Anna-Lena Schäfer, Maresa Weitmann, Petra Dinse, Emily Alberto, Michèle C Buck, Jonathan Jm Landry, Bianka Baying, Julia Slotta-Huspenina, Jenny Roesler, Patrick N Harter, Anne-Sophie Kubasch, Jörn Meinel, Eiman Elwakeel, Elisabeth Strack, Christine Tran Quang, Omar Abdel-Wahab, Marc Schmitz, Andreas Weigert, Tobias Schmid, Uwe Platzbecker, Vladimir Benes, Jacques Ghysdael, Halvard Bonig, Katharina S Götze, Carla V Rothlin, Sourav Ghosh, Hind Medyouf
ABSTRACT
Acute leukemias are systemic malignancies associated with a dire outcome. Because of low immunogenicity, leukemias display a remarkable ability to evade immune control and are often resistant to checkpoint blockade. Here, we discover that leukemia cells actively establish a suppressive environment to prevent immune attacks by co-opting a signaling axis that skews macrophages toward a tumor-promoting tissue repair phenotype, namely the GAS6/AXL axis. Using aggressive leukemia models, we demonstrate that ablation of the AXL receptor specifically in macrophages, or its ligand GAS6 in the environment, stimulates antileukemic immunity and elicits effective and lasting natural killer cell- and T cell-dependent immune response against naïve and treatment-resistant leukemia. Remarkably, AXL deficiency in macrophages also enables PD-1 checkpoint blockade in PD-1-refractory leukemias. Finally, we provide proof-of-concept that a clinical-grade AXL inhibitor can be used in combination with standard-of-care therapy to cure established leukemia, regardless of AXL expression in malignant cells. SIGNIFICANCE: Alternatively primed myeloid cells predict negative outcome in leukemia. By demonstrating that leukemia cells actively evade immune control by engaging AXL receptor tyrosine kinase in macrophages and promoting their alternative priming, we identified a target which blockade, using a clinical-grade inhibitor, is vital to unleashing the therapeutic potential of myeloid-centered immunotherapy.This article is highlighted in the In This Issue feature, p. 2659.
Sigma-Aldrich
Fluoromount Aqueous Mounting Medium, for use with fluorescent dye-stained tissues
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-GAS6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution