Merck
  • Nitrosonisoldipine is a selective inhibitor of inflammatory caspases and protects against pyroptosis and related septic shock.
Nitrosonisoldipine is a selective inhibitor of inflammatory caspases and protects against pyroptosis and related septic shock.
European journal of immunology (2021-01-18)
Qianyue Chen, Jiashuo Zheng, Dingyu Wang, Qiyao Liu, Lulu Kang, Xiang Gao, Zhaoyu Lin
ABSTRACT
Pyroptosis is a type of acute cell death that mainly occurs in immune cells. It is characterized with robust release of inflammatory cytokines and has emerged to play a critical role in the pathogenesis of sepsis-associated immune disorders. In this study, we screened for pyroptotic inhibitors with the ultimate goal to benefit sepsis treatments. Accidentally, we identified that nitrosonisoldipine (NTS), a photodegradation product of calcium channel inhibitor nisoldipine, inhibits noncanonical pyroptosis. Using murine immortalized BM-derived macrophage and human THP-1 cell line, we further discovered that NTS not only inhibits noncanonical pyroptosis mediated by caspase-11 or caspase-4 but also canonical pyroptosis mediated by caspase-1. Mechanistically, NTS directly inhibits the enzyme activities of these inflammatory caspases, and these inhibitory effects persist despite extensive washout of the drug. By contrast, apoptosis mediated by caspase-3/-7 was not affected by NTS. Mice pretreated with NTS intraperitoneally displayed improved survival rate and extended survival time in LPS- and polymicrobe-induced septic models, respectively. In conclusion, NTS is a selective inhibitor of inflammatory caspases that blocks both the noncanonical and canonical pyroptotic pathways. It is safe for intraperitoneal administration and might be used as a prototype to develop drugs for sepsis treatments.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)