Merck
  • SMYD2 targets RIPK1 and restricts TNF-induced apoptosis and necroptosis to support colon tumor growth.
SMYD2 targets RIPK1 and restricts TNF-induced apoptosis and necroptosis to support colon tumor growth.
Cell death & disease (2022-01-14)
Yu-Qiang Yu, Veronika Thonn, Jay V Patankar, Oana-Maria Thoma, Maximilian Waldner, Marta Zielinska, Li-Li Bao, Miguel Gonzalez-Acera, Stefan Wallmüller, Felix B Engel, Michael Stürzl, Markus F Neurath, Eva Liebing, Christoph Becker
ABSTRACT
SMYD2 is a histone methyltransferase, which methylates both histone H3K4 as well as a number of non-histone proteins. Dysregulation of SMYD2 has been associated with several diseases including cancer. In the present study, we investigated whether and how SMYD2 might contribute to colorectal cancer. Increased expression levels of SMYD2 were detected in human and murine colon tumor tissues compared to tumor-free tissues. SMYD2 deficiency in colonic tumor cells strongly decreased tumor growth in two independent experimental cancer models. On a molecular level, SMYD2 deficiency sensitized colonic tumor cells to TNF-induced apoptosis and necroptosis without affecting cell proliferation. Moreover, we found that SMYD2 targeted RIPK1 and inhibited the phosphorylation of RIPK1. Finally, in a translational approach, pharmacological inhibition of SMYD2 attenuated colonic tumor growth. Collectively, our data show that SMYD2 is crucial for colon tumor growth and inhibits TNF-induced apoptosis and necroptosis.
Product Description
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Anti-SMYD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture
Roche
Cytotoxicity Detection Kit (LDH), suitable for protein quantification, suitable for cell analysis, detection, sufficient for ≤2,000 tests