Merck
  • Adult medial habenula neurons require GDNF receptor GFRα1 for synaptic stability and function.
Adult medial habenula neurons require GDNF receptor GFRα1 for synaptic stability and function.
PLoS biology (2021-11-09)
Diana Fernández-Suárez, Favio A Krapacher, Katarzyna Pietrajtis, Annika Andersson, Lilian Kisiswa, Alvaro Carrier-Ruiz, Marco A Diana, Carlos F Ibáñez
ABSTRACT
The medial habenula (mHb) is an understudied small brain nucleus linking forebrain and midbrain structures controlling anxiety and fear behaviors. The mechanisms that maintain the structural and functional integrity of mHb neurons and their synapses remain unknown. Using spatiotemporally controlled Cre-mediated recombination in adult mice, we found that the glial cell-derived neurotrophic factor receptor alpha 1 (GFRα1) is required in adult mHb neurons for synaptic stability and function. mHb neurons express some of the highest levels of GFRα1 in the mouse brain, and acute ablation of GFRα1 results in loss of septohabenular and habenulointerpeduncular glutamatergic synapses, with the remaining synapses displaying reduced numbers of presynaptic vesicles. Chemo- and optogenetic studies in mice lacking GFRα1 revealed impaired circuit connectivity, reduced AMPA receptor postsynaptic currents, and abnormally low rectification index (R.I.) of AMPARs, suggesting reduced Ca2+ permeability. Further biochemical and proximity ligation assay (PLA) studies defined the presence of GluA1/GluA2 (Ca2+ impermeable) as well as GluA1/GluA4 (Ca2+ permeable) AMPAR complexes in mHb neurons, as well as clear differences in the levels and association of AMPAR subunits with mHb neurons lacking GFRα1. Finally, acute loss of GFRα1 in adult mHb neurons reduced anxiety-like behavior and potentiated context-based fear responses, phenocopying the effects of lesions to septal projections to the mHb. These results uncover an unexpected function for GFRα1 in the maintenance and function of adult glutamatergic synapses and reveal a potential new mechanism for regulating synaptic plasticity in the septohabenulointerpeduncular pathway and attuning of anxiety and fear behaviors.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Neural Cell Adhesion Molecule Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Glutamate receptor 1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Glutamate Receptor 4 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-GluR2 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Substance P Antibody, pain, clone NC1, culture supernatant, clone NC1, Chemicon®