- Development and validation of RP-HPLC method for determination of doxorubicin hydrochloride from vacuum foam dried formulation
Development and validation of RP-HPLC method for determination of doxorubicin hydrochloride from vacuum foam dried formulation
Research Journal of Pharmacy and Technology (2016)
Hajare AA, et al.