Merck
  • Photochemical Organocatalytic Synthesis of Thioethers from Aryl Chlorides and Alcohols.
Photochemical Organocatalytic Synthesis of Thioethers from Aryl Chlorides and Alcohols.
Journal of the American Chemical Society (2024-01-24)
Shuo Wu, Thomas Hin-Fung Wong, Paolo Righi, Paolo Melchiorre
ABSTRACT
Thioethers, often found in pharmaceuticals and natural compounds, typically involve metal cross-coupling reactions, high temperatures, and the use of disagreeable thiols for their synthesis. Here we present a straightforward, thiol-free organocatalytic protocol that uses mild conditions to stitch together inexpensive alcohols and aryl chlorides, yielding a diverse array of aryl alkyl thioethers. Central to this approach was the discovery that tetramethylthiourea can serve as a simple sulfur source upon intercepting photochemically generated aryl radicals. To form radicals, we used a readily available indole thiolate organocatalyst that, when excited with 405 nm light, gained a strongly reducing power, enabling the activation of typically unreactive aryl chlorides via single-electron transfer. Radical trapping by the thiourea, followed by an alcohol attack via a polar path, resulted in the formation of thioether products.
Sigma-Aldrich
1-Methylindoline-2-thione, ≥95%
Sigma-Aldrich
Indoline-2-thione, ≥-95%