Merck
  • A yeast display immunoprecipitation method for efficient isolation and characterization of antigens.
A yeast display immunoprecipitation method for efficient isolation and characterization of antigens.
Journal of immunological methods (2008-12-02)
Yong Ku Cho, Irene Chen, Xin Wei, Lingjun Li, Eric V Shusta
ABSTRACT
Yeast antibody display has found a wide variety of applications including antibody affinity maturation, epitope mapping, and library screening. Here we report a yeast display immunoprecipitation (YDIP) technique that employs yeast cells displaying single-chain antibody fragments (scFv) on their surface as affinity capture reagents to isolate and characterize antigens. We show that displayed single-chain antibody fragments are active in a variety of detergent solutions commonly used for immunoprecipitation and that the antigen-antibody interaction can be accurately quantified by YDIP coupled with flow cytometry. The YDIP method has also been optimized so that it is compatible with commonly used protein characterization tools such as Western blotting, silver staining, and mass spectrometry. From complex protein mixtures, we have used YDIP to isolate, analyze and sequence both soluble and plasma membrane antigens using tandem mass spectrometry. In the case of the membrane antigen, YDIP coupled with tandem mass spectrometry was successful in identifying neural cell adhesion molecule (NCAM) as the antigen for an antibody previously selected as binding to the plasma membranes of brain endothelial cells. The presented method therefore has potential to facilitate antibody-antigen characterization.
Sigma-Aldrich
Lysozyme from chicken egg white, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, 10 mg/mL
Sigma-Aldrich
Lysozyme from chicken egg white, dialyzed, lyophilized, powder, ~100000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, powder (crystalline), 70000-140000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Lysozyme from chicken egg white, aseptically filled
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Supelco
Lysozyme from chicken egg white, VETRANAL®, analytical standard
Sigma-Aldrich
Conalbumin from chicken egg white, BioReagent, suitable for cell culture
Sigma-Aldrich
Conalbumin from chicken egg white, Iron complex
Sigma-Aldrich
Conalbumin from chicken egg white, Substantially iron-free
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥90% (agarose gel electrophoresis)
Supelco
Albumin from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder
Sigma-Aldrich
Albumin from chicken egg white, powder, 62-88% (agarose gel electrophoresis)