Merck
  • Capture of heat-killed Mycobacterium bovis bacillus Calmette-Guérin by intelectin-1 deposited on cell surfaces.
Capture of heat-killed Mycobacterium bovis bacillus Calmette-Guérin by intelectin-1 deposited on cell surfaces.
Glycobiology (2009-01-31)
Shoutaro Tsuji, Makiko Yamashita, Donald R Hoffman, Akihito Nishiyama, Tsutomu Shinohara, Takashi Ohtsu, Yoshimi Shibata
ABSTRACT
Intelectin is an extracellular animal lectin found in chordata. Although human and mouse intelectin-1 recognize galactofuranosyl residues included in cell walls of various microorganisms, the physiological function of mammalian intelectin had been unclear. In this study, we found that human intelectin-1 was a serum protein and bound to Mycobacterium bovis bacillus Calmette-Guérin (BCG). Human intelectin-1-binding to BCG was inhibited by Ca(2+)-depletion, galactofuranosyl disaccharide, ribose, or xylose, and was dependent on the trimeric structure of human intelectin-1. Although monomeric, mouse intelectin-1 bound to BCG, with its C-terminal region contributing to efficient binding. Human intelectin-1-transfected cells not only secreted intelectin-1 into culture supernatant but also expressed intelectin-1 on the cell surface. The cell surface intelectin-1 was not a glycosylphosphatidylinositol-anchored membrane protein. Intelectin-1-transfected cells captured BCG more than untransfected cells, and the BCG adherence was inhibited by an inhibitory saccharide of intelectin-1. Intelectin-1-preincubated cells took up BCG more than untreated cells, but the adhesion of intelectin-1-bound BCG was the same as that of untreated BCG. Mouse macrophages phagocytosed BCG more efficiently in medium containing mouse intelectin-1 than in control medium. These results indicate that intelectin is a host defense lectin that assists phagocytic clearance of microorganisms.
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Supelco
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, Ultra pure