Merck
  • Tris buffer (THAM). An appraisal of its physiologic effects and clinical usefulness.
Tris buffer (THAM). An appraisal of its physiologic effects and clinical usefulness.
The New England journal of medicine (1966-04-07)
H L Bleich, W B Schwartz
ABSTRACT
It has been the purpose of this discussion to review critically the physiologic considerations and clinical evidence regarding the use of Tris buffer in the treatment of acid-base disorders. It is concluded that there is little current evidence, theoretical, experimental, or clinical, to support the view that Tris has a significant therapeutic role. Some possible exceptions to this general conclusion are considered.
Sigma-Aldrich
Trizma® hydrochloride, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)