Merck
  • Draft Whole-Genome Sequence of Bacillus sonorensis Strain L12, a Source of Nonribosomal Lipopeptides.
Draft Whole-Genome Sequence of Bacillus sonorensis Strain L12, a Source of Nonribosomal Lipopeptides.
Genome announcements (2013-03-30)
David B Adimpong, Kim I Sørensen, Dennis S Nielsen, Line Thorsen, Thomas B Rasmussen, Patrick M F Derkx, Lene Jespersen
ABSTRACT
The Bacillus sonorensis L12 draft genome sequence is approximately 4,647,754 bp in size with a G+C content of 45.2%. Over 86% of the genome contains protein-encoding genes, including several gene clusters for de novo biosynthesis of the nonribosomal lipopeptides iturin, bacitracin, and fengycin, which could mean that the strain exhibits antifungal effects.
Sigma-Aldrich
GenElute Bacterial Genomic DNA Kits, sufficient for 70 purifications
Pricing and availability is not currently available.