  • Search Results
  • Determination of resin acids during production of wood pellets--a comparison of HPLC/ESI-MS with the GC/FID MDHS 83/2 method.
Determination of resin acids during production of wood pellets--a comparison of HPLC/ESI-MS with the GC/FID MDHS 83/2 method.
Journal of environmental monitoring : JEM (2011-08-30)
Sara Axelsson, Kåre Eriksson, Ulrika Nilsson
ABSTRACT
Resin acids are constituents of natural and technical products of widespread use. Exposure is known to cause health effects in the airways and on the skin. Liquid chromatography/positive ion electrospray-mass spectrometry (HPLC/pos ESI-MS) was investigated for determination of 7-oxodehydroabietic (7-OXO), dehydroabietic (DHAA) and abietic acid (AA) in wood dust-containing air samples as a derivatisation-free alternative to the GC/FID HSE method 83/2, developed by the Health and Safety Executive UK. The resin acid 7-OXO was measured as a marker for oxidised resin acids, which are known to be the main contact allergens in colophonium. The found detection limits were 0.42 ng m(-3) for 7-OXO, 5.2 ng m(-3) for DHAA and 9.4 ng m(-3) for AA, respectively, which are considerably lower than with the GC/FID method (24, 115 and 89 ng m(-3)). The two methods correlated well, although consistently and significantly lower concentrations of 7-OXO were detected with LC/MS. The higher concentration of this compound with MDHS 83/2 is suggested to be an artefact from the derivatisation step in the presence of soluble wood dust remains.
Product Description
Supelco
Abietic acid, technical, ~75% (GC)
Sigma-Aldrich
Dehydroabietic acid, ≥95% (LC/MS-ELSD)