Merck
  • 3-(4-Aminobutyn-1-yl)pyridines: binding at alpha 4 beta 2 nicotinic cholinergic receptors.
3-(4-Aminobutyn-1-yl)pyridines: binding at alpha 4 beta 2 nicotinic cholinergic receptors.
Bioorganic & medicinal chemistry letters (2003-12-31)
Deniz Dogruer, Mase Lee, Malgorzata Dukat, M Imad Damaj, Billy R Martin, Richard A Glennon
ABSTRACT
The binding of a series pyridylbutynylamines 6 was examined at alpha4beta2 nACh receptors. Structural modifications, comparing 6 with pyridyl ethers 2, did not consistently result in parallel effects on receptor affinity, suggesting possible differences in their modes of binding. Furthermore, the binding of amine 6a seemed to be accounted for by the newer vector pharmacophore models.
Sigma-Aldrich
3-Butyn-1-ol, 97%
Pricing and availability is not currently available.