Merck
  • Indirect resolution of enantiomers of penicillamine by TLC and HPLC using Marfey's reagent and its variants.
Indirect resolution of enantiomers of penicillamine by TLC and HPLC using Marfey's reagent and its variants.
Biomedical chromatography : BMC (2007-05-23)
R Bhushan, H Brückner, Virender Kumar
ABSTRACT
TLC and HPLC methods were developed for indirect chiral separation of penicillamine (3,3-dimethylcysteine) enantiomers after derivatization with Marfey's reagent (FDNP-Ala-NH(2)) and two of its structural variants, FDNP-Phe-NH(2) and FDNP-Val-NH(2). The binary mobile phase of phenol-water (3:1 v/v) and solvent combinations of acetonitrile and triethylamine phosphate buffer were found to give the best separation in normal and reversed-phase TLC, respectively. The diastereomers were also resolved on a reversed-phase C18 HPLC column with gradient elution of acetonitrile and 0.01 m trifluoroacetic acid. The results due to these three reagents were compared. The method was successful for checking the enantiomeric impurity of l-penicillamine in d-penicillamine and to check the enantiomeric purity of pharmaceutical formulations of d-penicillamine. The method was validated for linearity, repeatability, limit of detection and limit of quantification.
Sigma-Aldrich
Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, powder
Supelco
Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, derivatization grade (chiral), LiChropur, ≥99.0%