Merck
  • The effects of 2-bromopalmitate on the fatty acid composition in differentiating adipocytes of red sea bream (Pagrus major).
The effects of 2-bromopalmitate on the fatty acid composition in differentiating adipocytes of red sea bream (Pagrus major).
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology (2009-01-27)
Hiromi Oku, Masaharu Tokuda, Tetsuya Umino
ABSTRACT
To determine whether external factors affect the adipogenic function of fish adipocytes, the effects of 2-bromopalmitate (a PPAR agonist) on the fatty acid composition in differentiating adipocytes of red sea bream were investigated in vitro. In the presence of 2-bromopalmitate, the red sea bream adipocytes were differentiated and the effects on the fatty acid composition and the adipogenic gene expression were analyzed. With the level of 2-bromopalmitate, the content of 16:1n-7, a delta-9 desaturation product, increased in association with the increase in a stearoyl CoA desaturase (SCD) gene expression level while the triglyceride accumulation was not affected. Subsequently, the effects on the bioconversion of the n-3 and n-6 fatty acids, which are main series of dietary essential fatty acids, were examined. In the presence of 300 microM of 18:3n-3 or 18:2n-6, red sea bream stromal-vascular cells accumulated the lipid in the cytoplasm within 3 days by the fatty acid uptake with the increase of corresponding fatty acid contents. Furthermore, in both the 18:3n-3 and 18:2n-6 stored cells, the products of delta-6 desaturation (18:4n-3 and 18:3n-6, respectively) and C(18-20) elongation (20:3n-3 and 20:2n-6, respectively) were detected. However, neither the delta-6 desatutration nor C(18-20) elongation of 18:3n-3 and 18:2n-6 were enhanced by 2-bromopalmitate treatment. In conclusion, the results indicate that the adipocyte function in fish, e.g. adipogenic gene expression and fatty acid composition, can be modified by external factors and a main effect of 2-bromopalmitate is the increase in the content of delta-9 desaturation product by stimulating the SCD gene expression.
Sigma-Aldrich
2-Bromohexadecanoic acid, ≥99.0% (GC)
Sigma-Aldrich
2-Bromohexadecanoic acid, ~97%