Merck
  • Biochemical analysis of a recombinant glutathione transferase from the cestode Echinococcus granulosus.
Biochemical analysis of a recombinant glutathione transferase from the cestode Echinococcus granulosus.
Acta tropica (2009-12-26)
Laura Harispe, Gabriela García, Paula Arbildi, Leticia Pascovich, Cora Chalar, Arnaldo Zaha, Cecilia Fernandez, Veronica Fernandez
ABSTRACT
Glutathione transferases (GSTs) are believed to be a major detoxification system in helminths. We describe the expression and functional analysis of EgGST, a cytosolic GST from Echinococcus granulosus, related to the Mu-class of mammalian enzymes. EgGST was produced as an enzymatically active dimeric protein (rEgGST), with highest specific activity towards the standard substrate 1-chloro-2,4-dinitrobenzene (CDNB; 2.5 micromol min(-1)mg(-1)), followed by ethacrynic acid. Interestingly, rEgGST displayed glutathione peroxidase activity (towards cumene hydroperoxide), and conjugated reactive carbonyls (trans-2-nonenal and trans,trans-2,4-decadienal), indicating that it may intercept damaging products of lipid peroxidation. In addition, classical GST inhibitors (cybacron blue, triphenylthin chloride and ellagic acid) and a number of anthelmintic drugs (mainly, hexachlorophene and rafoxanide) were found to interfere with glutathione-conjugation to CDNB; suggesting that they may bind to EgGST. Considered globally, the functional properties of rEgGST are similar to those of putative orthologs from Echinococcus multilcularis and Taenia solium, the other medically important cestodes. Interestingly, our results also indicate that differences exist between these closely related cestode GSTs, which probably reflect specific biological functions of the molecules in each parasitic organism.
Sigma-Aldrich
trans-2-Nonenal, 97%