Merck
  • Novel poly(amidoamine)s with pendant primary amines as highly efficient gene delivery vectors.
Novel poly(amidoamine)s with pendant primary amines as highly efficient gene delivery vectors.
Macromolecular bioscience (2010-01-30)
Min Liu, Jun Chen, Yan-Ping Cheng, Ya-Nan Xue, Ren-Xi Zhuo, Shi-Wen Huang
ABSTRACT
Three poly(amidoamine)s with pendant primary amines were synthesized by Michael polyaddition of a diamine to N,N-methylenebis(acrylamide). The poly(amidoamine)s with primary and tertiary amines possess a high buffer capacity between pH = 5 and 7, which facilitates the escape of polymer/DNA complexes from endosomes according to the 'proton sponge hypothesis'. In vitro cytotoxicity of poly(amidoamine)s depends on the side-chain structure of the polymer. Polymers 1a-1c can condense plasmid DNA by electrostatic interactions to form nanosized polyelectrolyte complexes with a positive surface charge. In vitro transfection results indicate that the transfection efficiency of polymer 1b under optimized conditions is comparable to that of commercial branched PEI 25,000.
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
N,N′-Methylenebisacrylamide solution, suitable for electrophoresis, 2% in H2O