Merck
Miniplate osteosynthesis with four different systems in sheep.
International journal of oral and maxillofacial surgery (2010-11-19)
R Gutwald, R Schön, M Metzger, K Kreutzer, B Rahn, R Schmelzeisen, S Sauerbier
ABSTRACT
The aim of this study was to compare a combination of a locking system with self-tapping (ST-L) or self-drilling-tapping (SDT-L) screws with a combination of conventional miniplates with self-tapping (ST) and self-forming (SF) screws. A standardized osteotomy and osteosynthesis with one of the above mentioned systems was performed in 24 sheep. Callus formation was measured with the help of CT scans assisted by a navigation system. Specimens of each osteotomy gap were taken and examined histologically. The best results were observed when self-tapping screws and the Mini-Locking-System (ST-L) were applied. The slowest healing was seen in animals treated with miniplates and SF screws. After 8 weeks an increase in bone formation could be observed in the ST, SF, SDT-L systems. The results after 8 weeks were comparable with those achieved by the ST-L system after 4 weeks. The improved stability of the osteosynthesis with the ST-L system resulted in early ossification of the osteotomy gap and the smallest amount of callus formation.
Product Description
Supelco
Xylenol Orange disodium salt, for spectrophotometric determination of metal ions
Pricing and availability is not currently available.