  • Search Results
  • Effect of gamma-irradiation on some chemical characteristics and volatile content of linseed.
Effect of gamma-irradiation on some chemical characteristics and volatile content of linseed.
Journal of medicinal food (2011-05-10)
Hasan Yalcin, Ismet Ozturk, Mehmet Hayta, Osman Sagdic, Tuncay Gumus
ABSTRACT
The effect of irradiation (2.5, 4.0, 5.5, and 7.0 kGy) on chemical properties and volatile contents of linseed was investigated. Consistent decreases were observed in both protein and oil content of the irradiated linseed samples with increasing irradiation doses. The ash content of the irradiated linseed samples increased significantly (P<.05) with increasing irradiation doses except for 5.5 kGy. Irradiation treatment caused irregular changes in palmitic and stearic acid content. Although styrene and p-xylene content decreased as a result of irradiation, 1-hexanol content only decreased at 7.0 kGy. Benzaldehyde, p-cymene, and nonanol were not determined at irradiation doses above 4.0 kGy.
Product Description
Supelco
1-Hexanol, analytical standard
Supelco
p-Xylene, analytical standard
Sigma-Aldrich
1-Hexanol, anhydrous, ≥99%
Sigma-Aldrich
p-Xylene, anhydrous, ≥99%
Sigma-Aldrich
1-Hexanol, ReagentPlus®, ≥99.5% (GC)
Sigma-Aldrich
Hexyl alcohol, natural, ≥98%, FCC, FG
Sigma-Aldrich
Hexyl alcohol, FCC, FG
Supelco
p-Xylene solution, certified reference material, 5000 μg/mL in methanol
Supelco
p-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
p-Xylene, suitable for HPLC, ≥99%
Sigma-Aldrich
p-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
1-Hexanol, reagent grade, 98%
Sigma-Aldrich
p-Xylene, ReagentPlus®, 99%