Merck
  • [Determination of avermectin, diclazuril, toltrazuril and metabolite residues in pork by high performance liquid chromatography-tandem mass spectrometry].
[Determination of avermectin, diclazuril, toltrazuril and metabolite residues in pork by high performance liquid chromatography-tandem mass spectrometry].
Se pu = Chinese journal of chromatography (2011-06-11)
Xiaoming Gong, Jun Sun, Jing Dong, Jinling Yu, Hongtao Wang
ABSTRACT
A method for the determination of avermectin, ivermectin, doramectin, moxidectin, eprinomectin, diclazuril, toltrazuril and its two metabolite residues in pork was developed using QuEChERS method with high performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS). The sample was extracted with acetonitrile and purified through QuEChERS method using ODS as the sorbent. The target compounds were separated on a Venusil ASB C18 column (150 mm x 2.1 mm, 3.0 microm) and detected by HPLC-MS/MS. The linear ranges were 0.005 - 0.2 mg/L and the correlation coefficients were all above 0.990. The average recoveries and the relative standard deviations ranged from 73.2% to 91.5% and from 12% to 17% at the spiked levels of 0.005, 0.01 and 0.02 mg/kg for the 9 analytes in pork matrix. This method is reliable, and suitable for the determination of the residues of avermectin and related compounds in pork.
Product Description
Diclazuril for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Diclazuril, VETRANAL®, analytical standard
Supelco
Toltrazuril, VETRANAL®, analytical standard