Merck
  • [Mechanisms of UV photodegradation on performance of a subsequent biofilter treating gaseous chlorobenzene].
[Mechanisms of UV photodegradation on performance of a subsequent biofilter treating gaseous chlorobenzene].
Huan jing ke xue= Huanjing kexue (2012-03-29)
Can Wang, Jin-Ying Xi, Hong-Ying Hu, Yuan Yao
ABSTRACT
To provide insight into effects of UV pretreatment on a subsequent biofilters, the mechanisms of UV pretreatment on biofilter performance were further studied. Results showed that gaseous chlorobenzene UV photodegradation products resulted in a pH decrease from 6-8 to 4-7 in filter bed. Ozone produced by UV photodegradation changed the physical properties of biofilm by reducing biofilm thickness and oxidizing surface part of the extra-cellular polymer substance, which promoted the oxygen, nutrient and pollutant transfer to biofilm. The reduction of biofilm thickness further resulted in the improvement of filter bed's physical properties by providing a higher specific surface area of 880 m x m(-3) compared with the value of 784 m2 x m(-3) without UV pretreatment.
Product Description
Supelco
Chlorobenzene, analytical standard
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Sigma-Aldrich
Chlorobenzene, suitable for HPLC, 99.9%
Sigma-Aldrich
Chlorobenzene, ReagentPlus®, 99%
Sigma-Aldrich
Chlorobenzene, ACS reagent, ≥99.5%