  • Search Results
  • Effect of dietary adipic acid and corn dried distillers grains with solubles on laying hen performance and nitrogen loss from stored excreta with or without sodium bisulfate.
Effect of dietary adipic acid and corn dried distillers grains with solubles on laying hen performance and nitrogen loss from stored excreta with or without sodium bisulfate.
Poultry science (2012-04-14)
C Romero, M E Abdallh, W Powers, R Angel, T J Applegate
ABSTRACT
Effects of dietary adipic acid (0 vs. 1%) and corn dried distillers grains with solubles (DDGS; 0 vs. 20%) were evaluated on hen performance and egg characteristics from 26 to 34 wk of age. Four isocaloric and isonitrogenous diets were randomly assigned to blocks of 6 consecutive cages (36 cages per diet; 2 hens per cage). On wk 2 and 7 of the experiment, excreta were collected by cage block, mixed, and equally split into 2 containers. Sodium bisulfate (SBS) was spread (8.8 kg/100 m(2)) on the top surface of half of the containers. All containers were stored uncovered for 14 d at room temperature. Excreta pH, DM, and N content were measured on d 0, 7, and 14 of storage. Feed intake (112 g/d per hen), egg production (96.1%), and egg specific gravity (1.079 g/g) were not affected by diet. On excreta collection day, a synergy (P = 0.014) between dietary adipic acid and DDGS was detected, as the lowest excreta pH was obtained with the diet including both adipic acid and DDGS. On d 7 of storage, excreta pH was still reduced by dietary adipic acid (P = 0.046) and DDGS (P < 0.001), but a week later, only dietary DDGS decreased excreta pH (8.91 vs. 9.21; P < 0.001). Whereas dietary adipic acid had no influence on excreta N loss, excreta from hens fed 20% DDGS lost 19.7% more N (P = 0.039) during storage than hens not eating DDGS. Surface amendment of excreta with SBS increased excreta DM content, with the effect being even more marked on d 14 of storage (increase of 6.7 percentage units; P < 0.001), consistently decreased excreta pH during storage (P < 0.001) and reduced N loss by 26.1% for the 14 d of storage period.
Sigma-Aldrich
Adipic acid, BioXtra, ≥99.5% (HPLC)
Sigma-Aldrich
Adipic acid, ≥99.5%
Sigma-Aldrich
Adipic acid, 99%
Supelco
Adipic acid, certified reference material, TraceCERT®