Merck
  • Bacteriorhodopsin: fourier transform infrared methods for studies of protonation of carboxyl groups.
Bacteriorhodopsin: fourier transform infrared methods for studies of protonation of carboxyl groups.
Methods in enzymology (1986-01-01)
G Dollinger, L Eisenstein, S L Lin, K Nakanishi, K Odashima, J Termini
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)