Merck
  • The syntheses of new camphorsulfonylated ligands derived from 2-amino-2'-hydroxy-1,1'-binaphthyl and their enantioselectivities in the addition of dialkylzinc reagents to aldehydes.
The syntheses of new camphorsulfonylated ligands derived from 2-amino-2'-hydroxy-1,1'-binaphthyl and their enantioselectivities in the addition of dialkylzinc reagents to aldehydes.
Chirality (2012-06-30)
Guangling Bian, Huayin Huang, Hua Zong, Ling Song
ABSTRACT
A series of new camphorsulfonylated ligands derived from chiral 2-amino-2'-hydroxy-1,1'-binaphthyl (NOBIN) and (+)-camphorsulfonic acid were synthesized by a short and simple synthetic sequence, and their enantioselective catalytic activities were assessed in the nucleophilic addition reaction of dialkylzinc reagents to aldehydes in the presence of titanium tetraisopropoxide. The most efficient ligand, N-hydroxycamphorsulfonylated (S)-NOBIN, gave (S)-addition products with good yields and up to 87% of ee value. The (1)H nuclear magnetic resonance (NMR) and (13)C NMR results of the titanium titration experiments on this ligand indicate that the most likely catalytic reactive species involved in this catalytic asymmetric addition is a bimetallic titanium complex. A possible catalytic reaction mechanism is proposed.
Sigma-Aldrich
D-Camphor, ≥97%, FG
Sigma-Aldrich
(1S)-(−)-Camphor, 95%
Supelco
D-Camphor, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(1R)-(+)-Camphor, 98%
Sigma-Aldrich
(±)-Camphor, ≥95.5%
Supelco
(−)-Camphor, analytical standard
Sigma-Aldrich
(±)-Camphor, purum, synthetic, ≥95.0% (GC)
Sigma-Aldrich
Camphor, 96%
Camphor (dl), primary reference standard
Sigma-Aldrich
(±)-Camphor, meets analytical specification of Ph. Eur., BP, ≥95% (GC), racemic
Camphor (racemic), European Pharmacopoeia (EP) Reference Standard