Merck
  • Competitive binding of abasic site-binding ligands and masking ligands to DNA duplexes for the analysis of single-base mutation.
Competitive binding of abasic site-binding ligands and masking ligands to DNA duplexes for the analysis of single-base mutation.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2013-01-11)
Yusuke Sato, Tomoe Kageyama, Seiichi Nishizawa, Norio Teramae
ABSTRACT
We describe a competitive binding assay for the analysis of single-base mutation by using abasic site (AP site)-specific binding fluorescent ligands and masking ligands bound to the AP site in DNA duplexes. 2-Amino-5,6,7-trimethyl-1,8-naphthyridine (ATMND) can strongly bind to not only cytosine (C), but also thymine (T) opposite an AP site in the DNA duplexes (K(11)/10(7) M(-1): C, 15; T, 7.0). By contrast, in the presence of lumichrome (Lch) as a masking ligand, ATMND shows a clear binding selectivity for C over T (K(11)/10(7) M(-1): C, 7.0; T, 0.57), which is ascribed to the effective masking effect of Lch to the ATMND-T interaction in the AP site-containing DNA duplexes. It was shown that the competitive binding of ATMND and Lch to the AP site allowed the highly selective detection of C-related single-base mutation in DNAs amplified by polymerase chain reaction.
Sigma-Aldrich
Lumichrome
Sigma-Aldrich
Thymine, ≥99%
Sigma-Aldrich
Thymine, BioReagent, suitable for cell culture