Merck
  • Core-shell nanocarriers with ZnO quantum dots-conjugated Au nanoparticle for tumor-targeted drug delivery.
Core-shell nanocarriers with ZnO quantum dots-conjugated Au nanoparticle for tumor-targeted drug delivery.
Carbohydrate polymers (2013-02-13)
Tong Chen, Tong Zhao, Dongfeng Wei, Yanxia Wei, Yuanyuan Li, Haixia Zhang
ABSTRACT
Core-shell structured multifunctional nanocarriers (NCs) of ZnO quantum dots-conjugated gold nanoparticles (Au NPs) as core and amphiphilic hyperbranched block copolymer as shell were synthesized for targeted anticancer drug delivery. The amphiphilic hyperbranched block copolymer contained poly(l-lactide) (PLA) inner arm and folate (FA)-conjugated a sulfated polysaccharide from Gynostemma pentaphyllum Makino (GPPS-FA) outer arm. The structure and properties of core-shell structured multifunctional nanocarriers were characterized and determined by UV-visible spectra, FT-IR spectra, X-ray diffraction (XRD), fluorescence spectroscopy and TEM analyses. The release results indicated that camptothecin (CPT) release from NCs at pH 7.4 was much greater than that at pH 5.3. The cytotoxicity studies showed that both the blank NCs and the CPT-loaded NCs provided high anticancer activity against Hela cells. Furthermore, nanocarriers gained specificity to target model cancer cells in this study due to the enhanced cell uptake mediated by FA moiety. The results indicated that the NCs not only had great potential as tumor-targeted drug delivery nanocarrier, but also had an assistant role in the treatment of cancer.
Sigma-Aldrich
Polylactic acid, Mw ~60,000
Pricing and availability is not currently available.