Merck
  • Characterization, pharmacokinetics, tissue distribution and antitumor activity of honokiol submicron lipid emulsions in tumor-burdened mice.
Characterization, pharmacokinetics, tissue distribution and antitumor activity of honokiol submicron lipid emulsions in tumor-burdened mice.
Die Pharmazie (2013-03-01)
Jiaxin Zheng, Yujie Tang, Moran Sun, Yuanyuan Zhao, Qiang Li, Jie Zhou, Yanzhi Wang
ABSTRACT
Honokiol, isolated from the Chinese traditional herb magnolia, is a poorly water-soluble component and has been found to have anti-tumor properties. In the current study, honokiol submicron lipid emulsions (HK-SLEs) were prepared by high pressure homogenization technology. After HK-SLEs were physically characterized, their pharmacokinetics, tissue distribution and antitumor activity after intravenous (i.v.) administration to tumor-burdened mice were examined, using honokiol solution (HK-SOL) as the control. The results showed that the mean particle size, zeta potential, pH value, osmolality, drug loading (DL)% and entrapment efficiency (EE)% of HK-SLEs were 186.6 +/- 1.7 nm, -35.65 +/- 0.67 mV, 7.22 +/- 0.26, 298 +/- 2.3 mOsm/L, 7.1 +/- 0.2% and 95.5 +/- 0.2%, respectively. HK-SLEs were stable for at least 12 months when stored at 4 +/- 2 degrees C. The pharmacokinetic results showed that the drug concentration-time curves of HK-SLEs and HK-SOL could both be described by an open two-compartment model. The half-life of HK-SLEs (t1/2(alpha) = 8.014 min, t1/2(beta) = 35.784 min) was remarkably prolonged compared to that of HK-SOL (t1/2(alpha) = 4.318 min, t1/2(beta) = 15.522 min). HK-SLEs exhibited a greater AUC and reduced plasma clearance. The tissue distribution results indicated that HK-SLEs have better targeting properties to lung and tumor tissues compared with those of HK-SOL. Both HK-SLEs and HK-SOL tended to accumulate in brain tissue. In vivo study showed that HK-SLEs treatment caused significant inhibition of mouse sarcoma S180 tumor growth compared to HK-SOL. These results suggest that HK-SLEs might be an effective parenteral carrier for honokiol delivery in cancer treatment.
Product Description
Supelco
Soybean oil, analytical standard
Sigma-Aldrich
Soybean oil, dietary source of long-chain triglycerides and other lipids
Sigma-Aldrich
Honokiol, ≥98% (HPLC), powder