Merck
  • Effectiveness and safety of lidocaine in the induction of fetal cardiac asystole for second trimester pregnany termination.
Effectiveness and safety of lidocaine in the induction of fetal cardiac asystole for second trimester pregnany termination.
Boletin de la Asociacion Medica de Puerto Rico (2013-06-19)
Ronald López-Cepero, Lauren Lynch, Alberto de la Vega
ABSTRACT
The presence of a major congenital anomaly is a frequent indication for late termination of pregnancy. The possibility of the fetus being born alive is significant, thus, feticide prior to the procedure is desirable. The purpose of this study was to assess the safety and efficacy of lidocaine 1% as a feticidal agent prior to second trimester termination of pregnancy. We conducted a chart review of all patients who underwent a second trimester termination of pregnancy at our institution between March 2009 and June 2012. We collected data regarding the indication for the termination procedure, gestational age, site of lidocaine injection, dosage of lidocaine, need for additional to produce asystole, and maternal complications. We identified 54 patients who underwent second trimester termination following injection with lidocaine. Forty-six cases (85%) were done for major fetal anomalies and 8 cases (15%) were for maternal indications. The mean gestational age was 22 weeks (SD = 2.3). The mean volume of lidocaine 1% injected was 10.1 mL (range: 5-40 mL). Asystole was achieved in 1-2 minutes following intracardiac administration. Intracardiac injection was successful in 45/46 (98%) of cases. Intrathoracic administration was successful in 5/6 (83%). This approach was chosen when cardiac puncture was not effective. Two fetuses receiving an initial intraabdominal or umbilical vein injection required additional doses of intracardiac lidocaine to produce asystole. There were no maternal complications. Intracardiac administration of lidocaine is an effective method to induce cardiac asystole for second trimester pregnancy termination. Extra-cardiac injection, however, is less effective.
Product Description
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Lidocaine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Lidocaine, European Pharmacopoeia (EP) Reference Standard
Lidocaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Lidocaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lidocaine hydrochloride monohydrate, solid
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Lidocaine, analytical standard