Merck
  • Application of a multidimensional gas chromatography system with simultaneous mass spectrometric and flame ionization detection to the analysis of sandalwood oil.
Application of a multidimensional gas chromatography system with simultaneous mass spectrometric and flame ionization detection to the analysis of sandalwood oil.
Journal of chromatography. A (2010-11-30)
Danilo Sciarrone, Rosaria Costa, Carla Ragonese, Peter Quinto Tranchida, Laura Tedone, Luca Santi, Paola Dugo, Giovanni Dugo, Daniel Joulain, Luigi Mondello
ABSTRACT
The production and trade of Indian sandalwood oil is strictly regulated, due to the impoverishment of the plantations; for such a reason, Australian sandalwood oil has been evaluated as a possible substitute of the Indian type. International directives report, for both the genuine essential oils, specific ranges for the sesquiterpene alcohols (santalols). In the present investigation, a multidimensional gas chromatographic system (MDGC), equipped with simultaneous flame ionization and mass spectrometric detection (FID/MS), has been successfully applied to the analysis of a series of sandalwood oils of different origin. A detailed description of the system utilized is reported. Three santalol isomers, (Z)-α-trans-bergamotol, (E,E)-farnesol, (Z)-nuciferol, epi-α-bisabolol and (Z)-lanceol have been quantified. LoD (MS) and LoQ (FID) values were determined for (E,E)-farnesol, used as representative of the oxygenated sesquiterpenic group, showing levels equal to 0.002% and 0.003%, respectively. A great advantage of the instrumental configuration herein discussed, is represented by the fact that identification and quantitation of target analytes are carried out in one step, without the need to perform two separate analyses.
Product Description
Supelco
3-Methyl-5-(2,2,3-trimethyl-3-cyclopenten-1-yl)-4-penten-2-ol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Sandalwood oil, natural
Pricing and availability is not currently available.