Merck
  • Validation of a computer case definition for sudden cardiac death in opioid users.
Validation of a computer case definition for sudden cardiac death in opioid users.
BMC research notes (2012-09-04)
Vivian K Kawai, Katherine T Murray, C Michael Stein, William O Cooper, David J Graham, Kathi Hall, Wayne A Ray
ABSTRACT
To facilitate the use of automated databases for studies of sudden cardiac death, we previously developed a computerized case definition that had a positive predictive value between 86% and 88%. However, the definition has not been specifically validated for prescription opioid users, for whom out-of-hospital overdose deaths may be difficult to distinguish from sudden cardiac death. We assembled a cohort of persons 30-74 years of age prescribed propoxyphene or hydrocodone who had no life-threatening non-cardiovascular illness, diagnosed drug abuse, residence in a nursing home in the past year, or hospital stay within the past 30 days. Medical records were sought for a sample of 140 cohort deaths within 30 days of a prescription fill meeting the computer case definition. Of the 140 sampled deaths, 81 were adjudicated; 73 (90%) were sudden cardiac deaths. Two deaths had possible opioid overdose; after removing these two the positive predictive value was 88%. These findings are consistent with our previous validation studies and suggest the computer case definition of sudden cardiac death is a useful tool for pharmacoepidemiologic studies of opioid analgesics.
Product Description
Supelco
(+)-Propoxyphene solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Hydrocodone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®