Merck
  • Relationship between the concentration of ethanol and methanol in blood samples from Swedish drinking drivers.
Relationship between the concentration of ethanol and methanol in blood samples from Swedish drinking drivers.
Forensic science international (1988-06-01)
A W Jones, H Löwinger
ABSTRACT
Headspace gas chromatography was used to determine the concentration of ethanol and methanol in blood samples from 519 individuals suspected of drinking and driving in Sweden where the legal alcohol limit is 0.50 mg/g in whole blood (11 mmol/l). The concentration of ethanol in blood ranged from 0.01 to 3.52 mg/g with a mean of 1.83 +/- 0.82 mg/g (+/- S.D.). The frequency distribution was symmetrical about the mean but deviated from normality. A plot of the same data on normal probability paper indicated that it might be composed of two subpopulations (bimodal). The concentration of methanol in the same blood specimens ranged from 1 to 23 mg/l with a mean of 7.3 +/- 3.6 mg/l (+/- S.D.) and this distribution was markedly skew (+). The concentration of ethanol (x) and methanol (y) were positively correlated (r = 0.47, P less than 0.001) and implies that 22% (r2) of the variance in blood-methanol can be attributed to its linear regression on blood-ethanol. The regression equation was y = 3.6 + 2.1 x and the standard error estimate was 0.32 mg/l. This large scatter precludes making reliable estimates of blood-methanol concentration from measurements of blood-ethanol concentration and the regression equation. But higher blood-methanol concentrations are definitely associated with higher blood-ethanol in this sample of Swedish drinking drivers. Frequent exposure to methanol and its toxic products of metabolism, formaldehyde and formic acid, might constitute an additional health risk associated with heavy drinking in predisposed individuals. The determination of methanol in blood of drinking drivers in addition to ethanol could indicate long-standing ethanol intoxication and therefore potential problem drinkers or alcoholics.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Progesterone, ≥99%
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Progesterone, VETRANAL®, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Supelco
5α-Dihydrotestosterone (DHT) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®